L(-)-Sorbose, Millipore®

Leverandør: Merck Millipore Calbiochem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: L-(-)-Sorbose, (3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one

567692-1 567692-250
567692-1EA 15340 DKK
567692-1 567692-250
L(-)-Sorbose, Millipore®
L(-)-Sorbose
Formel: C₆H₁₂O₆
Molekylvægt: 180.16 g/mol
Kogepunkt: 380
Smeltepunkt: 158…160 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00151097
CAS-nummer: 87-79-6
EINECS: 201-771-8
Merck Index: 12,08874

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR