Ethyl 6-Bromhexanoat 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-Bromocaproic acid ethyl ester, 6-Bromohexanoic acid ethyl ester, Ethyl 6-bromohexanoate
305920250
305921000
ACRO305920250EA
669
DKK
ACRO305920250
ACRO305921000
Ethyl 6-Bromhexanoat 98%
Ethyl 6-Bromhexanoat
Formel:
BrH₂C(CH₂)₄COOCH₂CH₃ Molekylvægt: 223.11 g/mol Kogepunkt: 128…130 °C (16 mmHg) Smeltepunkt: 33 Massefylde: 1.254 g/cm³ Flammepunkt: 58,3 °C |
MDL Number:
MFCD00000270 CAS-nummer: 25542-62-5 EINECS: 247-085-2 UN nummer: 3272 |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4580 to 1.4600 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...