4-Vinylphenylboronsyre 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dihydroxy(4-vinylphenyl)borane, (4-Vinylphenyl)metaboric acid, 4-Vinylbenzeneboronic acid

359810010 359810050
ACRO359810010EA 324 DKK
ACRO359810010 ACRO359810050
4-Vinylphenylboronsyre 98%
4-Vinylphenylboronsyre
Formel: C₈H₉BO₂
Molekylvægt: 147.97 g/mol
Smeltepunkt: 190 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00239441
CAS-nummer: 2156-04-9

Bestil nu

Specifikationer Testresultater

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR