5-fluorcytosin ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one
L16496.03 L16496.MD
L16496.03EA 1250 DKK
L16496.03 L16496.MD
5-fluorcytosin ≥98%
5-fluorcytosin
Purity: 98+% CAS No.: 2022-85-7 Molecular Formula: C4H4FN3O Formula Weight: 129.09 Melting Point: ca 297° dec. Beilstein Reference: 127285 Merck Reference: 14,4125 MDL No.: MFCD00006035
Formel: C₄H₄FN₃O
Molekylvægt: 129.09 g/mol
Kogepunkt: 298
Smeltepunkt: ∼ 297 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006035
CAS-nummer: 2022-85-7
EINECS: 217-968-7
Merck Index: 13,04151