Ammoniumjern(III)citrat, red brown
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate
A11199.30
A11199.0B
A11199.0I
A11199.30EA
352
DKK
A11199.30
A11199.0B
A11199.0I
Ammoniumjern(III)citrat, red brown
Ammoniumjern(III)citrat
CAS No.: 1185-57-5
Odour: Weak
Merck Reference: 14,4017
MDL No.: MFCD00013099
Formel:
C₆H₅+4y)Fe(x)N(y)O₇ Massefylde: 1.8 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00013099 CAS-nummer: 1185-57-5 EINECS: 214-686-6 Merck Index: 14,4017 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...