Nitrilotrieddikesyre 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: NTA, 2,2',2''-Nitrilotriacetic acid

415740010 415740050 415742500 415740051
ACRO415740010EA 2040 DKK
ACRO415740010 ACRO415740050 ACRO415742500 ACRO415740051
Nitrilotrieddikesyre 99%
Nitrilotrieddikesyre
Formel: C₆H₉NO₆
Molekylvægt: 191.14 g/mol
Kogepunkt: 498
Smeltepunkt: 245 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004287
CAS-nummer: 139-13-9
EINECS: 205-355-7
Merck Index: 12,06675

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration excess NaOH ≥98.5 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR