N,N-Diethylethylendiamin 98+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
150701000
150705000
ACRO150701000EA
329
DKK
ACRO150701000
ACRO150705000
N,N-Diethylethylendiamin 98+%
N,N-Diethylethylendiamin
Formel:
C₆H₁₆N₂ Kogepunkt: 146 °C (1013 hPa) Smeltepunkt: –70 °C Massefylde: 0,82 g/cm³ Flammepunkt: 30 °C |
MDL Number:
MFCD00008176 CAS-nummer: 100-36-7 EINECS: 202-844-7 UN nummer: 2685 ADR: 8,II |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4350 to 1.4370 (20°C, 589 nm) |
Water | ≤0.5 % (K.F.) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...