4-Nitro-2,6-xylenol ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 2-Hydroxy-5-nitro-3-xylene, 2,6-Dimethyl-4-nitrophenol
A15472.06 A15472.14
A15472.06EA 704 DKK
A15472.06 A15472.14
4-Nitro-2,6-xylenol ≥98%
4-Nitro-2,6-xylenol
Purity: 98% CAS No.: 2423-71-4 Molecular Formula: C8H9NO3 Formula Weight: 167.17 Melting Point: ca 168° dec. Beilstein Reference: 1873275
Formel: C₈H₉NO₃
Molekylvægt: 167.16 g/mol
Smeltepunkt: ∼ 168 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007339
CAS-nummer: 2423-71-4
EINECS: 219-353-9