ESD Safety Shoes, Lace-Up, Mattis XXSports Low S3
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720331/39 720331/40 720331/41 720331/42 720331/43 720331/44 720331/35 720331/36 720331/37 720331/38 720331/45 720331/46 720331/47 720331/48
ELTN720331/39EA 694 DKK
ELTN720331/39 ELTN720331/40 ELTN720331/41 ELTN720331/42 ELTN720331/43 ELTN720331/44 ELTN720331/35 ELTN720331/36 ELTN720331/37 ELTN720331/38 ELTN720331/45 ELTN720331/46 ELTN720331/47 ELTN720331/48
ESD Safety Shoes, Lace-Up, Mattis XXSports Low S3
Sko og støvler
ESD safety shoes made from hydrophobized textile material Cordura® with recycled material content and a TPU/PU sole (Wellmaxx Sports). The revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides, creating more energy in every step.
  • Breathable lining with recycled material content
  • Padded tongue
  • Full-length inlay sole SPORTIVE ESD black
  • Non-metallic, penetration-resistant midsolewith recycled material content
  • Steel toe cap
Suitable for use indoors and outdoors, in areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), and areas where there is a risk of electrostatic discharge (ESDS/ESD). Shoes are also suitable for workplaces on hard undergrounds.
Colour: Black/blue
Certificeringer: EN ISO 20345 S3 SRA, form A.