(4-fluorphenyl)acetylchlorid 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Fluorobenzeneacetyl chloride, 4-Fluorophenylacetyl Chloride

376120010 376120050 376120250
ACRO376120010EA 358 DKK
ACRO376120010 ACRO376120050 ACRO376120250
(4-fluorphenyl)acetylchlorid 98%
(4-fluorphenyl)acetylchlorid
Formel: (C₆H₄F)CH₂COCl
Kogepunkt: 60 °C (8 mmHg)
Massefylde: 1,259 g/cm³
Flammepunkt: 107 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00674181
CAS-nummer: 459-04-1
UN nummer: 3265
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5130 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR