Packo Biobased Jars
Leverandør: CURTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4305-80-004 4310-80-004
CURT4305-80-004EA 46740 DKK
CURT4305-80-004 CURT4310-80-004
Packo Biobased Jars
Prøvecontainere Allround kasser
The new Packo lid with integrated gasket prevents liquids from escaping or spilling out. Leak-tightness is tested by resting pots on the lid and side for a certain period of time thus burdening the gasket.
  • Quarter-turn screw lid saves handling time
  • Low moisture vapor transmission
  • Food safe
  • Tamper evident
Closing a Packo jar according to the manual is easy enough by hand. Closing and opening however can be made even easier and more ergonomic with a Lid Locker. This simple tool allows you to create more torque with less effort.
Certificeringer: UNMore Product Information