Defender™ 6000 Front Mount Bench Scales
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30608667 30608728 30608729 30608730 30608731 30608732 30612505 30612506 30612507 30612508 30612509 30612510
OHAU30608667EA 19250 DKK
OHAU30608667 OHAU30608728 OHAU30608729 OHAU30608730 OHAU30608731 OHAU30608732 OHAU30612505 OHAU30612506 OHAU30612507 OHAU30612508 OHAU30612509 OHAU30612510
Defender™ 6000 Front Mount Bench Scales
Vægte
Defender 6000 stands up to harsh cleaning in processing and packaging environments with IP68 immersion/ IP69K washdown protection, 316 stainless steel construction and hermetically-sealed load cell.
  • 6-digit, 7-segment LCD display with white backlight, 45 mm high digits
  • 6 D (LR20) Batteries (Included)
  • i-DT61PW indicator
Versatile scales include fast weight display and multiple software modes to meet food, chemical processing, and packaging needs.
150% overload capacity protection, 3-colour check light bar, menu and key lock, selectable environmental and auto-print settings, stability indicator, overload/underload indicators, low battery indicator, auto shut-off, auto tare, adjustable rubber feet.
316 stainless steel platform and frame, hermetically-sealed stainless steel load cell, IP68/IP69K food grade polycarbonate housing indicator and 316 stainless steel mounting bracket.
Weighing, counting with APW optimisation, percent weighing, checkweighing, checkcounting, dynamic weighing/display hold.
Certificeringer: NSF certified.