Defender™ 6000 Hybrid Bench Scales
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30575550 30575551 30575552 30575553 30575554 30575555 30612473 30612474 30612475 30612476 30612477 30612498
OHAU30575550EA 19250 DKK
OHAU30575550 OHAU30575551 OHAU30575552 OHAU30575553 OHAU30575554 OHAU30575555 OHAU30612473 OHAU30612474 OHAU30612475 OHAU30612476 OHAU30612477 OHAU30612498
Defender™ 6000 Hybrid Bench Scales
Vægte
Defender 6000 stands up to harsh cleaning in processing and packaging environments with IP68 immersion/ IP69K washdown protection, 316 stainless steel construction and hermetically-sealed load cell.
  • 6-digit, 7-segment LCD display with white backlight, 45 mm high digits
  • 6 D (LR20) Batteries (Included)
  • i-DT61PW indicator
Versatile scales include fast weight display and multiple software modes to meet food, chemical processing, and packaging needs.
150% overload capacity protection, 3-colour check light bar, menu and key lock, selectable environmental and auto-print settings, stability indicator, overload/underload indicators, low battery indicator, auto shut-off, auto tare, adjustable rubber feet.
316 stainless steel platform and frame, hermetically-sealed stainless steel load cell, IP68/IP69K food grade polycarbonate housing indicator and 316 stainless steel mounting bracket.
Weighing, counting with APW optimizsation, percent weighing, checkweighing, checkcounting,
dynamic weighing/display hold.
Certificeringer: NSF certified.