Height Endurance Mountaineer Helmets
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PS73ORR PS73RBR PS73RER PS73WHR PS73YER
PRTWPS73ORREA 413 DKK
PRTWPS73ORR PRTWPS73RBR PRTWPS73RER PRTWPS73WHR PRTWPS73YER
Height Endurance Mountaineer Helmets
Hjelme Sikkerhedshjelme
The Height Endurance Mountaineer helmet is the ultimate evolution in the Height Endurance range. This vented, peak-less helmet is designed for a variety of uses, and is ideal for working at height, rescue operations, and climbing activities.
  • Specially designed for working at height
  • Y-style, four-point chin strap
  • Size adjustable headband via wheel ratchet system
  • Inner expanded polystyrene foam
  • Hi-visibility colour for visibility in daylight
  • Light and robust
  • Compatible with PS58 helmet visor holder
  • Compatible with PW47 and PS47 clip-on ear muff
The inner EPS foam, is comfortable sweatband and wheel ratchet adjustment ensure optimal fit. Hi-visibility colours ensure visibility in daylight. A wide range of accessories are available.
Up to seven years lifetime from manufacturing date (as long as undamaged).
Certificeringer: EN 397 (impact/penetration/−30 °C). The inner EPS foam is fully certified to the EN 12492 standard.