Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (APC/iFluor® 700)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100321F0 100321F1 100321F2
AATB100321F0EA 2380 DKK
AATB100321F0 AATB100321F1 AATB100321F2
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (APC/iFluor® 700)
Antistoffer
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependents).
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Advarsel: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: APC/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human