Safety Waste Caps, S50
Leverandør: S.C.A.T.
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
108113 502031
SCTV108113EA 1030 DKK
SCTV108113 SCTV502031
Safety Waste Caps, S50
Låg Sikkerhedslåg og sikkerhedsaffaldslåg
SCAT Safety waste caps are hermetically sealing caps for the safe disposal of solvents and HPLC waste.
The SCAT exhaust filter with three layers of activated carbon binds vapours from solvents, acids and alkalis. This additionally ensures safe pressure equalisation and occupational safety.