Filter plates, 96-well, for aqueous filtration, AcroPrep™ Advance
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8119 8131 8031 8231 8127 8227 8151
738-0116EA 1920 DKK
738-0116 738-0119 518-0026 738-0130 738-0198 738-0199 738-0248
Filter plates, 96-well, for aqueous filtration, AcroPrep™ Advance
Mikroplader Filterplader
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications; gross fractionation and lysate clarification; and general filtration.
  • Effektiv partikler fjernes - en række membraner og pore størrelser der sikrer optimal behandling af