Salvburk, brunt glass
Leverandør: NORDIC PACK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
110500-160 110500-50STRUK
215-1808EA 41 DKK
215-1808 215-1804
Salvburk, brunt glass
Flasker Glasflasker
Glass, amber, with black screw cap.
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Amber coloured to protect light sensitive materials
Perfect for sample collection and storage. Ideal for paste like media and granular materials.