3-Hydroxy-p-anisidin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Hydroxy-4-anisidine, 5-Amino-2-methoxyphenol, 3-Hydroxy-4-methoxyaniline, 4-Amino-2-hydroxyanisole
H0634-25G
H0634-5G
TCIAH0634-25GEA
977
DKK
TCIAH0634-25G
TCIAH0634-5G
3-Hydroxy-p-anisidin ≥98.0% (by GC, titration analysis)
3-Hydroxy-p-anisidin
Formel:
C₇H₉NO₂ Molekylvægt: 139.15 g/mol Kogepunkt: 277 |
MDL Number:
MFCD00010222 CAS-nummer: 1687-53-2 EINECS: 216-873-8 |
|
Specifikationer Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 129.0 to 134.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...