(4-(Brommethyl)phenyl)eddikesyre ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-(Bromomethyl)phenylacetic acid

B1351-25G B1351-5G
TCIAB1351-25GEA 1090 DKK
TCIAB1351-25G TCIAB1351-5G
(4-(Brommethyl)phenyl)eddikesyre ≥98.0% (by titrimetric analysis)
(4-(Brommethyl)phenyl)eddikesyre
Formel: (C₆H₄(CH₂Br))CH₂COOH
MDL Number: MFCD00010632
CAS-nummer: 13737-36-5
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67785f9f1336c6a3313f556bf45aa97f6c31e1d&vid=c67785f9f1336c6a3313f556bf45aa97f6c31e1d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 178.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR