2-Bromthiazol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B1280-250G
B1280-25G
TCIAB1280-250GEA
3280
DKK
TCIAB1280-250G
TCIAB1280-25G
2-Bromthiazol ≥98.0% (by GC)
2-Bromothiazole
Formel:
C₃H₂BrNS Kogepunkt: 171°C Massefylde: 1,82 g/cm³ (20 °C) Flammepunkt: 63°C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00005316 CAS-nummer: 3034-53-5 EINECS: 221-229-4 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...