6-Aminonicotinonitril ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
6-Amino-3-pyridinecarbonitrile, 6-aminopyridine-3-carbonitrile, 2-Amino-5-cyanopyridine, 2-Aminopyridine-5-carbonitrile
A1642-25G
A1642-5G
TCIAA1642-25GEA
2730
DKK
TCIAA1642-25G
TCIAA1642-5G
6-Aminonicotinonitril ≥98.0% (by GC, titration analysis)
6-Aminonicotinonitril
Formel:
C₆H₅N₃ Kogepunkt: 250°C Smeltepunkt: 164 °C |
MDL Number:
MFCD00128902 CAS-nummer: 4214-73-7 UN nummer: 3439 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 161.0 to 165.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...