2-Mercapto-1-methylimidazol, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Mercapto-N-methyl-1H-imidazole, Thiamazole, Methimazole, 1-Methyl-1H-imidazole-2(3H)-thione, 2-Mercapto-N-methylimidazole, 2-Mercapto-1-methyl-1H-imidazole

M8506-100G M8506-25G
M8506-100GEA 1400 DKK
M8506-100G M8506-25G
2-Mercapto-1-methylimidazol, Supelco®
2-Mercapto-1-methylimidazole
Formel: C₄H₆N₂S
Kogepunkt: 155 °C (1013 hPa)
Massefylde: 1,36 g/cm³ (20 °C)
MDL Number: MFCD00179321
CAS-nummer: 60-56-0
EINECS: 200-482-4
Merck Index: 13,06000
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=449d7259e1b5500c794023e17d74ff462cbcb6e9&vid=449d7259e1b5500c794023e17d74ff462cbcb6e9&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=023ce39513505d75bdbd880746da2b4653bd852b&vid=023ce39513505d75bdbd880746da2b4653bd852b&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR