Castor olie

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi

4702.2 4702.1
ROTH4702.2EA 0 DKK
ROTH4702.2 ROTH4702.1
Castor olie
Castor olie
Kogepunkt: 313
Smeltepunkt: –10 °C
Massefylde: 0.95 g/cm³ (20 °C)
Flammepunkt: >113 °C (closed cup)
MDL Number: MFCD00130746
CAS-nummer: 8001-79-4
EINECS: 232-293-8
Merck Index: 13,01908

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR