Tricine
Leverandør: RESEARCH ORGANICS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
RES3077T-A103X
RES3077T-A102X
RESERES3077T-A103XEA
0
DKK
RESERES3077T-A103X
RESERES3077T-A102X
Tricine
Tricine
Formel:
(HOCH₂)₃CNHCH₂CO₂H Molekylvægt: 179.17 g/mol Smeltepunkt: 187 Massefylde: 1.523 g/cm³ (19.85 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004277 CAS-nummer: 5704-04-1 EINECS: 227-193-6 Merck Index: 13,09724 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...