(Chlormethyl)trimethylsilan, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
M-Clarity™ quality level = MQ200
8.18557.0025
8.18557.0100
8.18557.0025EA
1140
DKK
8.18557.0025
8.18557.0100
(Chlormethyl)trimethylsilan, Sigma-Aldrich®
(Chloromethyl)trimethylsilane
Formel:
ClH₂CSi(CH₃)₃ Kogepunkt: 98 °C (1013 hPa) Massefylde: 0,8861 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000878 CAS-nummer: 2344-80-1 EINECS: 219-058-5 UN nummer: 2985 ADR: 3 (8),II |
Specifikationer Testresultater
Solubility | (20 °C) insoluble |
Molar Mass | 122.66 g/mol |
Boiling Point | 97 - 98 °C |
Flash Point | 3 °C |
Refractive Index | 1.418 (20 °C, 589 nm) |
Density | 0.88 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...