2-Aminoanthracen 94%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L14269.03 L14269.06
L14269.03EA 455 DKK
L14269.03 L14269.06
2-Aminoanthracen 94%
2-Aminoanthracene

Purity: 94% CAS No.: 613-13-8 Molecular Formula: C14H11N Formula Weight: 193.25 Melting Point: 235-239° Beilstein Reference: 2209414

Formel: C₁₄H₁₁N
Kogepunkt: 371 °C (1013 hPa)
Smeltepunkt: 235…239 °C
Massefylde: >1 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003582
CAS-nummer: 613-13-8
EINECS: 210-330-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR