4,4'-Dibrombenzophenon ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Bis(4-bromophenyl) ketone
L08200.06 L08200.03
L08200.06EA 2740 DKK
L08200.06 L08200.03
4,4'-Dibrombenzophenon ≥98%
4,4'-Dibrombenzophenon
Purity: 98+% CAS No.: 3988-03-2 Molecular Formula: C13H8Br2O Formula Weight: 340.03 Melting Point: 172-176° Beilstein Reference: 2049958
Formel: C₁₃H₈Br₂O
Molekylvægt: 340.01 g/mol
Smeltepunkt: 172…176 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016330
CAS-nummer: 3988-03-2
EINECS: 223-632-0