2-Hexynoic acid 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L02526.03 L02526.06
L02526.03EA 327 DKK
L02526.03 L02526.06
2-Hexynoic acid 96%
2-Hexynoic acid

Purity: 96% CAS No.: 764-33-0 Molecular Formula: CH3(CH2)2C≡CCO2H Formula Weight: 112.13 Refractive Index: 1.4590 Melting Point: 23-26° Boiling Point: 110-112°/10mm Beilstein Reference: 1744983 UN No.: 3261 MDL No.: MFCD00014371

Formel: C₆H₈O₂
Kogepunkt: 110…112 °C (10 mmHg)
Smeltepunkt: 23…26 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014371
CAS-nummer: 764-33-0
UN nummer: 3261
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR