1-Methyl-3-phenyl-1-piperazin ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H26924.03 H26924.06 H26924.14
H26924.03EA 331 DKK
H26924.03 H26924.06 H26924.14
1-Methyl-3-phenyl-1-piperazin ≥97%
1-Methyl-3-phenyl-1-piperazin

Purity: 97% CAS No.: 5271-27-2 Molecular Formula: C11H16N2 Formula Weight: 176.26 Melting Point: 56-60° Boiling Point: 85°/0.5mm UN No.: 2811 MDL No.: MFCD03411603

Formel: C₁₁H₁₆N₂
Kogepunkt: 85 °C (0,5 mmHg)
Smeltepunkt: 56…60 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03411603
CAS-nummer: 5271-27-2
EINECS: 432-360-3
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR