N-Phenyl-p-phenylendiamin 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B23867.22 B23867.0B B23867.30
B23867.22EA 614 DKK
B23867.22 B23867.0B B23867.30
N-Phenyl-p-phenylendiamin 98%

Purity: 98% CAS No.: 101-54-2 Molecular Formula: C12H12N2 Formula Weight: 184.24 Flash Point: 193°(379°F) Melting Point: 72-76° Boiling Point: 354° Beilstein Reference: 908935 UN No.: 3077 MDL No.: MFCD00007850

Formel: C₁₂H₁₂N₂
Kogepunkt: 354 °C
Smeltepunkt: 72…76 °C
Massefylde: 1,09
Flammepunkt: 193 °C (379 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007850
CAS-nummer: 101-54-2
EINECS: 202-951-9
ADR: 9,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR