5-Methoxy-2-methylindol ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B21348.03 B21348.06
B21348.03EA 511 DKK
B21348.03 B21348.06
5-Methoxy-2-methylindol ≥99%
5-Methoxy-2-methylindole

Purity: 99+% CAS No.: 1076-74-0 Molecular Formula: C10H11NO Formula Weight: 161.20 Storage Temperature: 2°C to 8°C Melting Point: 85-88° MDL No.: MFCD00005620

Formel: C₁₀H₁₁NO
Kogepunkt: 145 °C (1,5 mmHg)
Smeltepunkt: 85…88 °C
MDL Number: MFCD00005620
CAS-nummer: 1076-74-0
EINECS: 214-066-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR