2'-Fluor-4'-methoxyacetophenon ≥99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
A16891.06 A16891.14 A16891.22
A16891.06EA 159 DKK
A16891.06 A16891.14 A16891.22
2'-Fluor-4'-methoxyacetophenon ≥99%
2'-Fluor-4'-methoxyacetophenon
Purity: 99% CAS No.: 74457-86-6 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Flash Point: 110°(230°F) Melting Point: 50-53° Boiling Point: 80-85°/0.1mm Beilstein Reference: 3539992 MDL No.: MFCD00042290
Formel: C₉H₉FO₂
Kogepunkt: 80…85 °C (0,1 mmHg)
Smeltepunkt: 50…53 °C
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042290
CAS-nummer: 74457-86-6