N,N,N-Trimethylaniliniumiodid ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N,N,N-Trimethylanilinium iodide, Trimethylphenylammonium iodide
A14402.14
A14402.22
A14402.36
A14402.14EA
401
DKK
A14402.14
A14402.22
A14402.36
N,N,N-Trimethylaniliniumiodid ≥99%
N,N,N-Trimethylaniliniumiodid
Purity: 99%
CAS No.: 98-04-4
Molecular Formula: C6H5N(CH3)3I
Formula Weight: 263.12
Melting Point: ca 227° subl.
Beilstein Reference: 3916752
Merck Reference: 14,7318
UN No.: 2811
MDL No.: MFCD00011791
Formel:
C₉H₁₄IN Smeltepunkt: ∼ 227 °C (subl.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011791 CAS-nummer: 98-04-4 EINECS: 202-630-3 UN nummer: 2811 ADR: 6.1,III Merck Index: 14,7318 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...