4,4'-Dichlorbenzophenon ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4-Dichlorobenzophenone, 4,4-DBP, Bis(4-chlorophenyl) ketone

A12608.14 A12608.22 A12608.36
A12608.14EA 503 DKK
A12608.14 A12608.22 A12608.36
4,4'-Dichlorbenzophenon ≥99%
4,4'-Dichlorbenzophenon

Purity: 99% CAS No.: 90-98-2 Molecular Formula: C13H8Cl2O Formula Weight: 251.11 Melting Point: 144-146° Boiling Point: 352-354° MDL No.: MFCD00000623

Formel: C₁₃H₈Cl₂O
Kogepunkt: 352…354 °C
Smeltepunkt: 144…146 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000623
CAS-nummer: 90-98-2
EINECS: 202-030-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR