4-Nitropyrocatechol ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Nitrocatechol, 4-Nitrobenzene-1,2-diol
A12122.06
A12122.14
A12122.22
A12122.06EA
574
DKK
A12122.06
A12122.14
A12122.22
4-Nitropyrocatechol ≥98%
4-Nitropyrocatechol
Purity: 98+%
CAS No.: 3316-09-4
Molecular Formula: C6H5NO4
Formula Weight: 155.11
Melting Point: 174-177°
Beilstein Reference: 1867508
Formel:
C₆H₅NO₄ Smeltepunkt: 174…177 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007242 CAS-nummer: 3316-09-4 EINECS: 222-009-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...