4-Propylcyclohexanon ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A11126.14
A11126.22
A11126.14EA
1540
DKK
A11126.14
A11126.22
4-Propylcyclohexanon ≥99%
4-Propylcyclohexanone
Purity: 99%
CAS No.: 40649-36-3
Molecular Formula: C9H16O
Formula Weight: 140.23
Flash Point: 86°(186°F)
Refractive Index: 1.4535
Boiling Point: 115°/36mm
Beilstein Reference: 2076023
MDL No.: MFCD00058690
Formel:
C₉H₁₆O Kogepunkt: 115 °C (36 mmHg) Massefylde: 0,910 Flammepunkt: 86 °C (186 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00058690 CAS-nummer: 40649-36-3 EINECS: 406-810-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...