Levulinic acid ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Oxovaleric acid, 4-Ketovaleric acid, 4-Oxopentanoic acid

A10813.0B A10813.0I A10813.30
A10813.0BEA 1450 DKK
A10813.0B A10813.0I A10813.30
Levulinic acid ≥98%
Levulinic acid

Purity: 98% CAS No.: 123-76-2 Molecular Formula: C5H8O3 Formula Weight: 116.12 Flash Point: 137°(278°F) Refractive Index: 1.4415 Odour: Characteristic pH: 2.5-3.0 Melting Point: 26-31° Boiling Point: 245-246° Beilstein Reference: 506796 Merck Reference: 14,5472 UN No.: 3261 MDL No.: MFCD00002796

Formel: C₅H₈O₃
Kogepunkt: 245…246 °C
Smeltepunkt: 26…31 °C
Massefylde: 1,134
Flammepunkt: 137 °C (278 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002796
CAS-nummer: 123-76-2
EINECS: 204-649-2
UN nummer: 3261
ADR: 8,III
Merck Index: 14,5472

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR