Methylbromacetat ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Bromoacetic methyl ester

A10605.0B A10605.18 A10605.30
A10605.0BEA 3350 DKK
A10605.0B A10605.18 A10605.30
Methylbromacetat ≥98%
Methylbromacetat

Purity: 98+% CAS No.: 96-32-2 Molecular Formula: C3H5BrO2 Formula Weight: 152.98 Flash Point: 62°(143°F) Refractive Index: 1.4585 Odour: Pungent Melting Point: -50° Boiling Point: 144-145° Beilstein Reference: 506256 UN No.: 2643 MDL No.: MFCD00000189

Formel: BrCH₂COOCH₃
Kogepunkt: 144…145 °C
Smeltepunkt: –50 °C
Massefylde: 1,655
Flammepunkt: 62 °C (143 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000189
CAS-nummer: 96-32-2
EINECS: 202-499-2
UN nummer: 2643
ADR: 6.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR