3-Phenylpropanal 95%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Phenylpropionaldehyde, Hydrocinnamaldehyde
Purity: 95%
CAS No.: 104-53-0
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 95°(203°F)
Physical Form: Liquid
Density: 1.017
Refractive Index: 1.5230
Sensitivity: Air Sensitive
Appearance: Colourless
Melting Point: -42°
Boiling Point: 222-223°
Beilstein Reference: 1071910
MDL No.: MFCD00007021
Formel:
C₉H₁₀O Molekylvægt: 134.18 g/mol Kogepunkt: 222…223 °C Smeltepunkt: –42 °C Massefylde: 1.017 Flammepunkt: 95 °C (203 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007021 CAS-nummer: 104-53-0 EINECS: 203-211-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...