2-Brom-5-(trifluormethyl)pyridin ≥96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
43381.03
43381.06
43381.03EA
1040
DKK
43381.03
43381.06
2-Brom-5-(trifluormethyl)pyridin ≥96%
2-Bromo-5-(trifluoromethyl)pyridine
Purity: 96%
CAS No.: 50488-42-1
Molecular Formula: C6H3BrF3N
Formula Weight: 226.00
Physical Form: Crystalline
Melting Point: 45-47°
Boiling Point: 78-81°/30mm
MDL No.: MFCD00153086
Formel:
C₆H₃BrF₃N Kogepunkt: 78…81 °C (30 mmHg) Smeltepunkt: 45…47 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00153086 CAS-nummer: 50488-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...