Formaldehyd 37% i vandigopløsning stabilised ACS
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Stabilization:
stabiliseret med methanol 10-15 %
CAS No.: 50-00-0
Molecular Formula: CH2O
Formula Weight: 30.03
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 1.083
Refractive Index: 1.3765
Appearance: Colourless
Odour: Pungent
Melting Point: -15°
Boiling Point: 97°
Beilstein Reference: 1209228
Merck Reference: 14,4235
UN No.: 1198
MDL No.: MFCD00003274
Formel:
CH₂O Kogepunkt: 97 °C Smeltepunkt: –15 °C Massefylde: 1,083 Flammepunkt: 60 °C (140 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003274 CAS-nummer: 50-00-0 EINECS: 200-001-8 UN nummer: 1198 ADR: 3,III Merck Index: 14,4235 |