Ursodeoxygaldesyre 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
208590010
208590050
208590250
ACRO208590010EA
257
DKK
ACRO208590010
ACRO208590050
ACRO208590250
Ursodeoxygaldesyre 99%
Ursodeoxygaldesyre
Formel:
C₂₄H₄₀O₄ Kogepunkt: 547 °C (1013 hPa) Smeltepunkt: 203…206 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003680 CAS-nummer: 128-13-2 EINECS: 204-879-3 Merck Index: 12,10026 |
Specifikationer Testresultater
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Melting point | 200 to 206 °C |
Specific optical rotation | +58° to +62° (20 °C, 589 nm) (c=2, ethanol) |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...