m-Toluonitril 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: m-Tolunitrile, m-Methylbenzonitrile, 3-Methylbenzonitrile

158010050 158011000 158015000
ACRO158010050EA 283 DKK
ACRO158010050 ACRO158011000 ACRO158015000
m-Toluonitril 99%
m-Toluonitril
Formel: C₈H₇N
Kogepunkt: 99…101 °C (20 mmHg)
Smeltepunkt: –23 °C
Massefylde: 0,97 g/cm³
Flammepunkt: 86 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001808
CAS-nummer: 620-22-4
EINECS: 210-631-5

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5246 to 1.5266 (20°C, 589 nm)
GC ≥98.5 %
UV at 275 nm A: ≤0.1
Color scale ≤10 APHA
Stabilizer 2.5 ppm BHT
Residue after evaporation ≤0.01 %
Peroxides ≤0.005 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR