2-Aminothiazol 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Thiazol-2-ylamine, Aminothiazole, Thiazol-2-amine

153891000 153895000
ACRO153891000EA 217 DKK
ACRO153891000 ACRO153895000
2-Aminothiazol 97%
2-Aminothiazole
Formel: C₃H₄N₂S
Kogepunkt: 117 °C (20mbar)
Smeltepunkt: 86…91 °C
Massefylde: 1,5 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005325
CAS-nummer: 96-50-4
EINECS: 202-511-6
Merck Index: 13,00479

Bestil nu

Specifikationer Testresultater

Appearance Brown Powder or flakes or granules and/or lumps
Infrared spectrum Conforms
Melting point 86 to 91 °C
Water ≤0.75 % (K.F.)
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR