Methacrylsyreamid 98%, extra pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

149202500 149205000 149200025
ACRO149202500EA 279 DKK
ACRO149202500 ACRO149205000 ACRO149200025
Methacrylsyreamid 98%, extra pure
Methacrylsyreamid
Formel: C₄H₇NO
Molekylvægt: 85.11 g/mol
Kogepunkt: 215 °C (760 mmHg)
Smeltepunkt: 108…112 °C
Flammepunkt: 103
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008018
CAS-nummer: 79-39-0
EINECS: 201-202-3

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Crystals
Infrared spectrum Conforms
Melting point 108 to 112 °C
Water ≤1 % (K.F.)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR