Trimethylorthoformat 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
148660010
148661000
148665000
148660050
ACRO148660010EA
615
DKK
ACRO148660010
ACRO148661000
ACRO148665000
ACRO148660050
Trimethylorthoformat 99%
Trimethylorthoformat
Formel:
C₄H₁₀O₃ Kogepunkt: 101…102 °C (760 mmHg) Smeltepunkt: –53 °C Massefylde: 0,97 g/cm³ Flammepunkt: 13 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008483 CAS-nummer: 149-73-5 EINECS: 205-745-7 UN nummer: 3271 ADR: 3,II Merck Index: 12,07012 |
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3780 to 1.3800 (20°C, 589 nm) |
GC | ≥98.5 % |
Elemental Analysis | 8.51 to 9.21 % (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...