(±)-Tetrahydrofurfurylalkohol 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
(±)-2-Hydroxymethyloxolan
138180010
138180025
ACRO138180010EA
419
DKK
ACRO138180010
ACRO138180025
(±)-Tetrahydrofurfurylalkohol 98%
(±)-Tetrahydrofurfurylalkohol
Formel:
C₅H₁₀O₂ Molekylvægt: 102.13 g/mol Kogepunkt: 178 Smeltepunkt: –80 °C Massefylde: 1.0543 g/cm³ Flammepunkt: 74 °C (closed cup) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005372 CAS-nummer: 97-99-4 EINECS: 202-625-6 Merck Index: 12,09353 |
Specifikationer Testresultater
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4510 to 1.4530 (20°C, 589 nm) |
Water | ≤0.3 % |
GC | ≥97.5 % |
Color scale | ≤50 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...