Phthalaldehyd 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

131080050 131080250 131081000 131085000
ACRO131080050EA 585 DKK
ACRO131080050 ACRO131080250 ACRO131081000 ACRO131085000
Phthalaldehyd 98+%
Phthalaldehyd
Formel: C₈H₆O₂
Kogepunkt: 83…84 °C (1hPa)
Smeltepunkt: 54…57 °C
Massefylde: 1,13 g/cm³ (20 °C)
Flammepunkt: 132 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00003335
CAS-nummer: 643-79-8
EINECS: 211-402-2
UN nummer: 2923
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Yellow Powder or crystals and/or lumps
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR