4-Nitropyrocatechol 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Nitrocatechol, 4-Nitrobenzene-1,2-diol

128600050 128600100 128600500
ACRO128600050EA 429 DKK
ACRO128600050 ACRO128600100 ACRO128600500
4-Nitropyrocatechol 97%
4-Nitropyrocatechol
Formel: C₆H₅NO₄
Smeltepunkt: 173…177 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007242
CAS-nummer: 3316-09-4
EINECS: 222-009-0

Bestil nu

Specifikationer Testresultater

Appearance Yellow to yellow-green to yellow brown Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 177 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR